missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | Sr(NO3)2 |
UN Number | 1507 |
IUPAC Name | strontium;dinitrate |
Formula Weight | 211.63g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Packaging | Poly bottle |
Chemical Name or Material | Strontium Nitrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'