missing translation for 'onlineSavingsMsg'
Learn More
Strontium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10254400

Strontium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10254400
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10254400

Brand: Fisher Chemical S/8400/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 645°C
CAS 10042-76-9
Color White
pH 5.0 to 7.0
Physical Form Crystalline Solid
Quantity 500 g
Molecular Formula N2O6Sr
MDL Number MFCD00011248
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name strontium;dinitrate
Molecular Weight (g/mol) 211.63
PubChem CID 24848
Formula Weight 211.63g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Strontium nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.