missing translation for 'onlineSavingsMsg'
Learn More
Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric), Honeywell Fluka™ Product Code.: 15663480

Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric), Honeywell Fluka™

Product Code. 15663480 Shop All Honeywell Fluka Products
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 15663480

Brand: Honeywell-Fluka 31633250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (complexometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 10042-76-9
Quantity 250 g
Molecular Formula N2O6Sr
Linear Formula Sr(NO3)2
MDL Number MFCD00011248
UN Number 1507
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
IUPAC Name strontium;dinitrate
Molecular Weight (g/mol) 211.63
PubChem CID 24848
Formula Weight 211.63g/mol
Percent Purity ≥99% (complexometric)
Grade ACS/puriss. p.a.
Chemical Name or Material Strontium Nitrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.