missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxide, tech., Thermo Scientific Chemicals

£102.51 - £1351.73

Chemical Identifiers

CAS 1314-11-0
Molecular Formula OSr
Molecular Weight (g/mol) 103.619
MDL Number MFCD00049558
InChI Key UFQXGXDIJMBKTC-UHFFFAOYSA-N
PubChem CID 518709
IUPAC Name oxostrontium
SMILES O=[Sr]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321839
View Documents
Thermo Scientific Alfa Aesar
012495.30
250 g
£102.51
250g
Please to purchase this item. Need a web account? Register with us today!
11311839
View Documents
Thermo Scientific Alfa Aesar
012495.A1
1 kg
£307.53
1kg
Please to purchase this item. Need a web account? Register with us today!
11301839
View Documents
Thermo Scientific Alfa Aesar
012495.A7
5 kg
£1351.73
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In the manufacture of strontium salts, pyrotechnics, pigments, in greases and soaps and as a desiccant.Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industries. It is an active component of paint dryers. It is also used to prepare pure strontium upon heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
Specifications

Chemical Identifiers

1314-11-0
103.619
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
OSr
MFCD00049558
518709
O=[Sr]

Specifications

∼3000°C
1314-11-0
OSr
UN3262
Soluble in fused potassium hydroxide; Insoluble in ether and acetone; Decomposes in water to Sr(OH)2 with evolution of heat;Slightly soluble in alcohol; Insoluble in acetone and ether.
O=[Sr]
103.619
103.62
Odorless
4.700 g/mL
2430°C
Powder
MFCD00049558
14,8846
UFQXGXDIJMBKTC-UHFFFAOYSA-N
oxostrontium
518709
Technical
Moisture sensitive
Strontium oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 215-219-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only