missing translation for 'onlineSavingsMsg'
Learn More

Strontium perchlorate trihydrate, 98%

Product Code. 15482377
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 15482377

Brand: Thermo Scientific Alfa Aesar 045577.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15650-09-6 | Cl2H6O11Sr | 340.56 g/mol

Strontium perchlorate trihydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15650-09-6
Molecular Formula Cl2H6O11Sr
Molecular Weight (g/mol) 340.56
MDL Number MFCD09039117
InChI Key VRRVWIQWZWFYDJ-UHFFFAOYSA-L
Synonym strontium perchlorate trihydrate, 2clo4.sr.3h2o, strontium perchlorate, trihydrate, strontium 2+ trihydrate diperchlorate
PubChem CID 71435311
SMILES O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98%
Solubility Information Very soluble in water. Soluble in alcohol.
IUPAC Name strontium;diperchlorate;trihydrate
Formula Weight 340.54 (286.52 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Strontium perchlorate trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.