missing translation for 'onlineSavingsMsg'
Learn More

Strontium peroxide, 12.3% available oxygen

Product Code. 15442647
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 15442647

Brand: Thermo Scientific Alfa Aesar 046310.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-18-7 | O2Sr | 119.618 g/mol

Applications of Strontium Peroxide include pyrotechnics i.e. fireworks (red color), ammunitions (tracer/hunting), oxidizer, alarm rocket. It is also used in textiles for fibers bleaching and as an oxidizing agent for hair bleaching (professional).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-18-7
Molecular Formula O2Sr
Molecular Weight (g/mol) 119.618
MDL Number MFCD00084686
InChI Key UHCGLDSRFKGERO-UHFFFAOYSA-N
Synonym strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygen
PubChem CID 14807
IUPAC Name strontium;peroxide
SMILES [O-][O-].[Sr+2]

Specifications

Melting Point 215°C (decomposition)
Color Brown
Physical Form Powder
Quantity 250 g
Linear Formula SrO2
UN Number UN1509
Merck Index 14,8847
Solubility Information Insoluble in water and slowly decomposes in water.
Formula Weight 119.62
Sensitivity Moisture sensitive
Chemical Name or Material Strontium peroxide, 12.3% available oxygen
Show More Show Less
compliance-icons
Product Identifier
  • Strontium peroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Strontium peroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.