missing translation for 'onlineSavingsMsg'
Learn More

Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11301459
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Sr(NO3)2 in 5% HNO3
MDL Number MFCD00134060

Specifications

Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Strontium, plasma standard solution, Sr 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'