missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£70.00 - £103.00
Chemical Identifiers
CAS | 7440-24-6 |
---|---|
Molecular Formula | Sr |
Molecular Weight (g/mol) | 87.62 |
InChI Key | CIOAGBVUUVVLOB-UHFFFAOYSA-N |
PubChem CID | 5359327 |
ChEBI | CHEBI:33324 |
IUPAC Name | strontium |
SMILES | [Sr] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12962195
|
Fisher Chemical
A 043 100 |
100 mL | Bottle |
£73.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12972195
|
Fisher Chemical
A 043 250 |
250 mL | Bottle |
£70.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12982195
|
Fisher Chemical
A 043 500 |
500 mL | Bottle |
£103.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7440-24-6 | |
87.62 | |
5359327 | |
strontium |
Sr | |
CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
CHEBI:33324 | |
[Sr] |
Specifications
1g/L | |
Single-component Standard Solution | |
CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
strontium | |
5359327 | |
H2O | |
Strontium |
7440-24-6 | |
Sr | |
[Sr] | |
87.62 | |
CHEBI:33324 | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)