missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Product Code. 11397597
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7759-02-6
Molecular Formula O4SSr
Molecular Weight (g/mol) 183.68
MDL Number MFCD00036274
InChI Key UBXAKNTVXQMEAG-UHFFFAOYSA-L
Synonym strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g
PubChem CID 3084026
IUPAC Name strontium;sulfate
SMILES [O-]S(=O)(=O)[O-].[Sr+2]

Specifications

Melting Point 1600°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula SrSO4
Merck Index 14,8848
Formula Weight 183.68
Percent Purity 99.9965%
Grade Puratronic™
Odor Odorless
Density 3.960 g/mL
Assay (metals basis)
Chemical Name or Material Strontium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'