missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11308827
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7759-02-6

Specifications

Melting Point 1600°C
Physical Form Crystalline
Quantity 100 g
Molecular Formula O4SSr
MDL Number MFCD00036274
Merck Index 14,8848
Synonym strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g
Solubility Information Soluble in dilute hydrochloric acid and nitric acid . Slightly soluble in water < 0.01g/L . Insoluble in ethanol and alkalis.
InChI Key UBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Sr+2]
IUPAC Name strontium;sulfate
Molecular Weight (g/mol) 183.68
PubChem CID 3084026
Formula Weight 183.68
Grade Reagent
Odor Odorless
Density 3.960 g/mL
Chemical Name or Material Strontium sulfate, For reagent grade
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'