missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfide, 99.9% (metals basis)

Product Code. 11483850
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11483850

Brand: Thermo Scientific Alfa Aesar 089895.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-96-1 | SSr | 119.68 g/mol

Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-96-1
Molecular Formula SSr
Molecular Weight (g/mol) 119.68
MDL Number MFCD00049560
InChI Key ZEGFMFQPWDMMEP-UHFFFAOYSA-N
Synonym strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
PubChem CID 14820
IUPAC Name strontium(2+) sulfanediide
SMILES [S--].[Sr++]

Specifications

Melting Point >2000°C
Color Gray
Physical Form Powder
Quantity 10 g
Merck Index 14,8849
Solubility Information Slightly soluble in water; Decomposes in acids
Formula Weight 119.68
Percent Purity 99.9%
Odor Like rotten eggs
Sensitivity Hygroscopic
Density 3.70 g/mL
Assay (metals basis)
Chemical Name or Material Strontium sulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Strontium sulphide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Strontium sulphide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.