missing translation for 'onlineSavingsMsg'
Learn More

Strontium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

Used in electronics and in electrical insulation

£41.51 - £106.53

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341199
View Documents
Thermo Scientific Alfa Aesar
011399.30
250 g
£41.51
250g
Please to purchase this item. Need a web account? Register with us today!
11351199
View Documents
Thermo Scientific Alfa Aesar
011399.A1
1 kg
£106.53
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Strontium titanium oxide is used as a diamond stimulant and also used as a strontium-90 in radioisotope thermoelectric generators. It is used in the electronics and electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12060-59-2
183.48
VEALVRVVWBQVSL-UHFFFAOYSA-N
82899
[Sr++].[O-][Ti]([O-])=O
O3SrTi
MFCD00049554
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
strontium;dioxido(oxo)titanium

Specifications

2060°C
Powder
MFCD00049554
Insoluble in water and most solvents.
[Sr++].[O-][Ti]([O-])=O
183.48
183.51
Odorless
(metals basis)
12060-59-2
O3SrTi
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium;dioxido(oxo)titanium
82899
≥99%
4.810 g/mL
Strontium titanium oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-044-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only