missing translation for 'onlineSavingsMsg'
Learn More

Strontium titanium oxide, S.A. 12.5-25m2/g, Thermo Scientific Chemicals

Catalog Number 11378768
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
12060-59-2
O3SrTi
183.48
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[Sr++].[O-][Ti]([O-])=O
This item is not returnable. View return policy
12060-59-2
O3SrTi
183.48
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
82899
strontium;dioxido(oxo)titanium
[Sr++].[O-][Ti]([O-])=O

Strontium titaniumoxide is used as paint . It has been used as a strontium-90-containing material in radioisotope thermoelectric generators. It is also used as a diamond simulant. This compound is been used in television sets, radios and computers, capacitors, because of the dielectric constant of the material. Used in electronics and in electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2060°C
Physical Form Powder
Quantity 100 g
Solubility Information It is insoluble in water and most solvents.
Formula Weight 183.51
Odor Odorless
Density 4.810 g/mL
Chemical Name or Material Strontium titanium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'