missing translation for 'onlineSavingsMsg'
Learn More

Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished Two Sides, 100 Orientation, Thermo Scientific™

100 Orientation

£232.00 - £924.00

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
SMILES [Sr++].[O-][Ti]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364158
View Documents
Thermo Scientific Alfa Aesar
041440.KT
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11374158
View Documents
Thermo Scientific Alfa Aesar
041440.KU
5 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12060-59-2
183.48
VEALVRVVWBQVSL-UHFFFAOYSA-N
82899
O3SrTi
MFCD00049554
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
[Sr++].[O-][Ti]([O-])=O

Specifications

12060-59-2
Substrate
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium;dioxido(oxo)titanium
82899
Strontium Titanium Oxide Substrate, Polished Two Sides
White
O3SrTi
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
[Sr++].[O-][Ti]([O-])=O
183.48
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-044-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications