missing translation for 'onlineSavingsMsg'
Learn More

Strontium titanium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Product Code. 11348857
1 Ea.
Click to view available options
Quantity:
1 Ea.
5 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

Strontium titanium oxide substrate is used as diamond stimulant, in precision optics, in varistors, and in advanced ceramics. It is also used in in radioisotope thermoelectric generators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Color White
Physical Form Substrate
Quantity 1 Ea.
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium titanium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.