Learn More
Strontium titanium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
£239.19 - £1232.13
Chemical Identifiers
CAS | 12060-59-2 |
---|---|
Molecular Formula | O3SrTi |
Molecular Weight (g/mol) | 183.48 |
MDL Number | MFCD00049554 |
InChI Key | VEALVRVVWBQVSL-UHFFFAOYSA-N |
Synonym | strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder |
PubChem CID | 82899 |
IUPAC Name | strontium;dioxido(oxo)titanium |
SMILES | [Sr++].[O-][Ti]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368857
|
Thermo Scientific Alfa Aesar
038507.KT |
1 Ea. |
£239.19
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358857
|
Thermo Scientific Alfa Aesar
038507.KU |
5 Ea. |
£1232.13
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12060-59-2 | |
183.48 | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
82899 | |
[Sr++].[O-][Ti]([O-])=O |
O3SrTi | |
MFCD00049554 | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
strontium;dioxido(oxo)titanium |
Specifications
12060-59-2 | |
Substrate | |
MFCD00049554 | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
strontium;dioxido(oxo)titanium | |
82899 | |
Strontium Titanium Oxide Substrate, Polished One Side |
White | |
O3SrTi | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
[Sr++].[O-][Ti]([O-])=O | |
183.48 | |
Odorless |
Safety and Handling
missing translation for 'einecsNumber' : 235-044-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.