missing translation for 'onlineSavingsMsg'
Learn More

Strontium tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

£87.33 - £308.54

Chemical Identifiers

CAS 13451-05-3
MDL Number MFCD00049562
Synonym Strontium tungstate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314877
View Documents
Thermo Scientific Alfa Aesar
041899.14
25 g
£87.33
25g
Please to purchase this item. Need a web account? Register with us today!
11304877
View Documents
Thermo Scientific Alfa Aesar
041899.22
100 g
£308.54
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13451-05-3
MFCD00049562
Strontium tungstate

Specifications

13451-05-3
Powder
MFCD00049562
335.46
Odorless
(metals basis)
White
SrWO4
Strontium tungstate
99.9%
6.187 g/mL
Strontium tungsten oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H335
May cause respiratory irritation.

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 236-617-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only