missing translation for 'onlineSavingsMsg'
Learn More
Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals Product Code.: 11321719

Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals

Product Code. 11321719
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11321719

Brand: Thermo Scientific Alfa Aesar 012346.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Mostly as intermediates( primary and secondary ). For chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-39-4
Molecular Formula O3SrZr
Molecular Weight (g/mol) 226.84
MDL Number MFCD00049555
InChI Key FCCTVDGKMTZSPU-UHFFFAOYSA-N
Synonym strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olateShow MoreShow Less
PubChem CID 82852
IUPAC Name strontium;dioxido(oxo)zirconium
SMILES [Sr++].[O-][Zr]([O-])=O

Specifications

Boiling Point 9000°C
Melting Point 2700°C
Color White
Physical Form Powder
Quantity 250 g
Formula Weight 226.84
Percent Purity 99.3%
Odor Odorless
Density 5.5 g/mL
Assay (metals basis)
Chemical Name or Material Strontium zirconium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE30Y031 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012346.30,012346.A1
Lot NumberC21Q31 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012346.30,012346.A1
Lot NumberW15A014 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012346.30,012346.A1
Lot NumberE23X027 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012346.30,012346.A1
Lot NumberS25A035 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012346.30,012346.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.