Learn More
Cole-Parmer™ Stuart™ Analogue Tube Roller
Provides a gentle, but highly efficient, rocking and rolling action. Stuart™ Analogue Tube Roller Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C.
Brand:  Cole-Parmer™ 04653-04
 View more versions of this product
Product Code. 11416498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Provides a gentle, but highly efficient, rocking and rolling action. The six rollers design has a small space-saving footprint, while the nine roller unit has a higher capacity.
  • Control is through a simple on/off switch and the unit will run continuously at the fixed speed of 33rpm
  • Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required.
  • Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C. Both roller mixers are robustly constructed and designed for easy cleaning, having plastic rollers and a drip tray to collect accidental spillages.
  • Most sizes of tubes, Bijoux, Universals and bottles can be accommodated, up to 100mm diameter.
Specifications
Specifications
33 rpm
230 V
50 Hz
110 mm
595 mm
SRT9
Up to 80%
1 Ea.
230 V, 50 Hz, 50 W
50 W
375 mm
340 mm
Analogue
Mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required
9 Rollers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Stuart™ Analogue Tube Roller > Capacity: 9Roller