Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Use these blocks for a variety of applications. Stuart™ Anodized Aluminum Blocks may be used in any combination.
£133.00 - £189.00
Product Code | Brand | Diameter (Metric) Well | Depth (Metric) Well | No. of Places | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Well | Depth (Metric) Well | No. of Places | Price | Quantity & Availability | |||
11496588
|
Cole-Parmer™
36610-59 |
30 mm | 47 mm | 4 |
£157.00
Each |
In Stock
Log in to see stock. |
|||
11486588
|
Cole-Parmer™
36610-56 |
25.5 mm | 47 mm | 6 |
£155.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||
11426598
|
Cole-Parmer™
36610-57 |
28 mm | 47 mm | 6 |
£155.00
Each |
In Stock
Log in to see stock. |
|||
11416588
|
Cole-Parmer™
36610-51 |
19.5 mm | 47 mm | 8 |
£155.00
Each |
In Stock
Log in to see stock. |
|||
11426588
|
Cole-Parmer™
36610-52 |
10.5 mm | 33 mm | 20 |
£155.00
Each |
In Stock
Log in to see stock. |
|||
11476578
|
Cole-Parmer™
36610-44 |
10 mm | 47 mm | 20 |
£133.00
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
12088493
|
Cole-Parmer™
36610-46 |
12.6 mm | 33 mm | 20 |
£164.00
Each |
In Stock
Log in to see stock. |
|||
11496578
|
Cole-Parmer™
SHT1/13 |
13.5 mm | 47 mm | 20 |
£155.00
Each |
In Stock
Log in to see stock. |
|||
11406588
|
Cole-Parmer™
SHT1/16 |
16.5 mm | 47 mm | 12 |
£154.00
Each |
In Stock
Log in to see stock. |
|||
11446588
|
Cole-Parmer™
36610-54 |
10.7 mm | 14 mm | 20 |
£154.00
Each |
In Stock
Log in to see stock. |
|||
11456588
|
Cole-Parmer™
36610-58 |
7.8 mm | 14 mm | 30 |
£189.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||
11466588
|
Cole-Parmer™
36610-62 |
61 mm | 14 mm | 48 |
£185.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||
Description
- For use with Stuart block heaters
- Manufactured from anodized aluminum
- Separate hole accommodates a thermometer if desired
Specifications
Anodized Aluminum | |
Block |