Learn More
Cole-Parmer™ Dry Block Heater Cover
Click to view available options
For Use With (Equipment):
2-block System
3-block System or Dual Control Heater
Description
- The heater covers ensure temperatures are maintained in your dry block heaters
- Model 36610-96 cover is suitable for use with two block heaters
- Model 36610-97 is suitable for use with three block or dual control heaters
- The extraction tool, 36610-98, makes removing blocks from the heater quick and effortless
Specifications
Specifications
For Use With (Equipment) | 2-block System |
Type | Safety Cover |