Learn More
Cole-Parmer™ Drum for DR-200 Series Rotating Shaker System Drives
Optimize performance of your DR-200 Series Rotating Shaker System with interchangeable drums.
Brand: Cole-Parmer™ 51600-97
This item is not returnable.
View return policy
Description
- Change your rotator's capacity with interchangeable drums
- Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Drum | |
For changing your rotator's capacity | |
One Bottle, Figure-8 Motion | |
1 |