Learn More
Cole-Parmer™ WS-100 Series Stuart™ Water Still
Save money while maintaining high performance with this WS-100 Series Stuart Water Still.
Brand:  Cole-Parmer™ 99293-00
 View more versions of this product
Product Code. 13145990
  • £1315.00 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Two independent thermostats prevent overheating.
  • Features a sturdy metal stand for either wall or bench mounting.
  • Operate directly from tap water.
  • This basic economical water still is perfect for general laboratory and educational distillations.
  • Borosilicate glass boiler and condenser provide the highest purity distillate.
  • The unique condenser design ensures that the distilled water droplets remain in contact with the cooling coil for an extended period for maximum energy transfer and immediate cold distilled water.
  • Condenser with screw thread connections and plastic 3/8 in. ID hose barb fittings allow quick and safe tubing change without worrying about breaking the glassware.
  • Easily descale without dismantling the glassware using the built-in acid addition funnel and wastewater drain.
  • Long-life chromium-plated metal heater is protected by twin safety thermostats.
Specifications
Specifications
Merit Water Still
220/240 V 50/60 Hz 3 kW
1
25°C to 35°C
5.9 in.
17.7 in.
19.7 in.
4 L/hr.
50/60 Hz
240 V
W4000
150 mm
450 mm
500 mm