missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ Heater Block Inserts

Product Code. 15103037
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Description:
Large Base
Medium Base
Small and Medium Bases
Quantity:
1/Ea.
3/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Accessory heating block for 100 mL round bottom flasks for use with the Stuart™ SHB1000BASE and Stuart™ Undergrad hotplate

  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks

Specifications

Description Medium Base
Capacity (Metric) 250 mL
Type Block
Quantity 3/Pk.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'