Learn More
Cole-Parmer™ Stuart™ Heater Block Insert
Click to view available options
Tube Size:
Medium
Large
Description
- Safest way to heat round-bottom flasks
- Flexible modular system for a wide range of sizes up to 500 mL
- Low-well design is perfect for multi-neck flasks
Specifications
Specifications
Material | Anodized Aluminum |
Type | Block |
Includes | base SHB250BASE |
Tube Size | Medium |