missing translation for 'onlineSavingsMsg'
Learn More

Bibby Scientific™ Stuart™ PCR Plate Dri Block Insert with Thermometer Hole

Product Code. 11882380
Click to view available options
No. of Places:
96
This item is not returnable. View return policy
This item is not returnable. View return policy

Block heaters provide a dry, constant temperature source in the laboratory. These 96-well PCR insert blocks are made of aluminium alloy and must be ordered separately from the heater units.

  • 96-Well PCR block insert plate
  • Hole diameter 5.5 mm
  • Hole depth 17.3 mm
  • Tapered base profile
  • Aluminium alloy
  • Must be ordered separately from the heater unit

Specifications

Description PCR Plate Dri-Block Insert
For Use With (Equipment) Dry Block Heater
Material Anodized Aluminum
No. of Places 96
Type Insert Block
For Use With (Application) Constant, dry temperature - incubation, boiling, inactivation, wet ashing, sample concentration, enzyme analysis and many other clinical industrial purposes
Includes 96 Holes, 17.3 mm Deep Tapered, PCR Plate, With Thermometer Hole
Dimensions (L x W x H) 95 x 150 x 61 mm
Width (English) 5.9 in.
Width (Metric) 150 mm
Height (English) 2.4 in.
Height (Metric) 61 mm
Depth (English) 3.7 in.
Depth (English) Well 0.7 in.
Depth (Metric) 95 mm
Depth (Metric) Well 17.3 mm
Diameter (English) Well 0.2 in.
Diameter (Metric) Well 5.5 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.