Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Optimize performance of your DR-200 Series Rotating Shaker System with this Clip Plate for Rotator Drums.
Brand:  Cole-Parmer™ 51600-91
 View more versions of this product
Product Code. 12033324
  • £191.00 / Pack of 2
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
Tube Rack
12
12 mm
Tubes (12 mm)
For 12 Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Clip Plate for Rotator Drum > Clip Plate for Rotator Drum; For use with: 12 mm Tubes