Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Optimize performance of your DR-200 Series Rotating Shaker System with this Clip Plate for Rotator Drums.
Brand:  Cole-Parmer™ 51600-94
 View more versions of this product
Product Code. 12063324
  • £181.00 / Pack of 2
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
Tube Rack
7
24 mm
Tubes (24 mm)
For 7 Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Clip Plate for Rotator Drum > Clip Plate for Rotator Drum; For use with: 24 mm Tubes