Learn More
Cole-Parmer™ Stuart™ SCT1 Temperature Controller
Designed for use with the Stuart™ Undergrad range of stirrer hotplates, the SCT1 can be used either as a temperature controller or as a digital thermometer. Stuart™ SCT1 Temperature Controller is ideal for the accurate temperature control of aqueous or oil based samples in the laboratory.
Brand:  Cole-Parmer™ 04805-92
Product Code. 12008323
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Accurate liquid temperature control
  • Built in retort fitting and probe holder
  • Detachable temperature probe
  • Bright, easy to read LED display
  • Compact and light weight
  • Comprehensive range of accessories
Specifications
Specifications
±0.5°C
90 mm
PT100
123 mm
SCT1