Learn More
Cole-Parmer™ INC-200 Series Stuart™ Hybridization Incubator/Shaker
INC-200 Series Stuart™ Hybridization Incubator/Shaker is versatile—features both rotisserie and rocking motions.
Brand:  Cole-Parmer™ 39454-05
Product Code. 11496558
  • £3920.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Forced air circulation combined with microprocessor control result in excellent temperature uniformity.
  • Up-and-over door for easy access to the chamber.
  • Use a hybridization incubator when it is vital to maintain the integrity of the sample while heating it to the exact temperature.
  • The incubator is ideal for nucleic acid hybridization applications such as Northern (RNA) blots, Southern (DNA) blots, and Western (protein) blots.
  • This compact incubator comes with two built-in shakers.
  • Rotisserie shaker has a rotational speed from 2 to 10 rpm while the platform shaker has a rocking speed from 5 to 70 oscillations/minute.
  • The gentle rocking action is ideal for the washing stage of most hybridization procedures.
  • Safety features include an overtemperature protection system, plus the power is cut to the motor if the motor overheats or stalls.
  • Stainless steel interior is easy to clean.
  • Double-glazed smoked acrylic and polycarbonate door provides radiation protection.
  • Order a rotisserie that accepts 75 mm OD bottles and hybridization bottles separately.
  • Bottles are made from borosilicate glass and are autoclavable.
Specifications
Specifications
20 L
Hybridization Oven