missing translation for 'onlineSavingsMsg'
Learn More
Styrene, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals Product Code.: 10627362

Styrene, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals

Product Code. 10627362
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10627362

Brand: Thermo Scientific Acros 220532500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5, 98-29-3
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name styrene
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31.0°C
Density 0.9060g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CH2
Refractive Index 1.5460 to 1.5480
Quantity 250 mL
Beilstein 05, 474
Merck Index 15, 8990
Specific Gravity 0.906
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers
Stabilizer 10 to 20ppm p-tert-butylcatechol
Viscosity 0.695 mPa.s (25°C)
Formula Weight 104.15
Percent Purity 99.5%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Styrene, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463010 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number220530010,220532500
Lot NumberA0463010 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number220530010,220532500
Lot NumberA0457938 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number220530010,220532500
Lot NumberA0458889 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog Number220530010,220532500
Lot NumberA0454404 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number220530010,220532500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Styrene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Styrene, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.