missing translation for 'onlineSavingsMsg'
Learn More

Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals

Product Code. 10398093
Click to view available options
Quantity:
10 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5, 98-29-3
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name styrene
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31.0°C
Density 0.9060g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Linear Formula C6H5CH=CH2
Refractive Index 1.5460 to 1.5480
Quantity 25 L
Beilstein 05, 474
Merck Index 15, 8990
Specific Gravity 0.906
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers
Stabilizer 10 to 20ppm p-tert-butylcatechol
Viscosity 0.695 mPa.s (25°C)
Formula Weight 104.15
Vapor Pressure 7mbar at 20°C
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Styrene, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Styrene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'