missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Styrene Wall Thermometer

Brannan™ Styrene Wall Thermometer is large with bold black print and suitable for indoor and outdoor use.

Brand:  Brannan™ 14/433/3

Product Code. 13152201

  • £13.95 / Each
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Large (380mm)
  • Wall thermometer
  • Bold black print
  • Indoor or outdoor use
  • Celsius and Faherenheit scales
Specifications

Specifications

Styrene
Wall Thermometer
-20°C
±1.0°C
74 g
50°C
-20°C to +50°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers