missing translation for 'onlineSavingsMsg'
Learn More

SU 5416, Tocris Bioscience™

Product Code. 18024962
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

VEGFR inhibitor. Also inhibits KIT, RET, MET and FLT3

Inhibitor of vascular endothelial growth factor receptor (VEGFR) that also inhibits other tyrosine kinases KIT, MET, FLT3 and RET. Displays no activity against EGFR, HER2, IGF1R and PDGFR. Inhibits tumor vascularization and growth of multiple tumor types.

Chemical Identifiers

CAS 204005-46-9
Molecular Formula C15H14N2O
Molecular Weight (g/mol) 238.29
InChI Key WUWDLXZGHZSWQZ-WQLSENKSSA-N
Synonym semaxanib, semaxinib, semoxind, semaxanib su5416, semaxnib, tsu 16, unii-71ia9s35aj, vegf receptor 2 kinase inhibitor iii, tsu-16, semaxanib usan/inn
PubChem CID 5329098
IUPAC Name (3Z)-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylidene]-1H-indol-2-one
SMILES CC1=CC(=C(N1)C=C2C3=CC=CC=C3NC2=O)C

Specifications

Quantity 10 mg
Formula Weight 238.28
Percent Purity >99%
Chemical Name or Material SU 5416
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'