missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SU 5416, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of vascular endothelial growth factor receptor (VEGFR) that also inhibits other tyrosine kinases KIT, MET, FLT3 and RET. Displays no activity against EGFR, HER2, IGF1R and PDGFR. Inhibits tumor vascularization and growth of multiple tumor types.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 238.28 |
Percent Purity | >99% |
Chemical Name or Material | SU 5416 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'