missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SUB-GEL Teaching Horizontal Gel Tank Package

SUB-GEL Teaching gel tanks are ideal for demonstration of horizontal gel electrophoresis, helps teachers to demonstrate horizontal gel electrophoresis to many students using just single gel tank and power supply.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 15351357

  • £773.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • High quality injection moulded construction
  • Leak free ‘Plug-and-Go’ casting dams
  • UV transparent gel trays
  • Height adjustable combs
  • Low gel and buffer volumes
  • Cassette-style electrodes - difficult to break and easy to change
  • Each tank is available with or without power supply
Specifications

Specifications

Standard combs: Eight x 8 sample, 1mm MC (2 combs per gel)
Demonstrating Horizontal Gel Electrophoresis to Students
7 x 7 cm (2.7 x 2.7 in.)
64 sample with standard comb option, 128 samples using optional 16 sample combs
26.5 x 17.5 x 9 cm (10.4 x 6.8 x 3.5 in.)
Maximum Gel throughput: Four 70mm x 70mm mini Gels
Teaching Gel Tank, 4 UV trays
Four 70mm x 70mm UV trays, eight 8 sample combs, eight casting dams, loading guides and cables; Plus power supply (Cat. No 12643546)
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers