missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUB6 Submarine Gel Electrophoresis Unit PROMO

Buy any Hoefer™ horizontal electrophoresis apparatus and get a 25g bottle of Agarose (HBGR14025)

Brand:  Hoefer™ SUB10

Code : A0

Additional Details : Weight : 0.10000kg

Product Code. 11514494

  • £477.98 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Maximum 80-sample throughput capacity
  • Features removable gel casting tray with end gaskets that allow gels to be cast directly within tank
  • Ideal for routine preparatory and analytical electrophoresis techniques
Specifications

Specifications

EN61010-1, CE
80%
80
45°C
9.05 x 6.4 in. (23 x 16.5 cm)
3.9 in.
10 cm
4.5 in.
11.5 cm
300 V
200 mA
Mini Plus Submarine Gel Electrophoresis Unit
Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips
113°F
0.45 L
9.05 in.
23 cm
6.4 in.
16.5 cm
6.5 cm
60 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers