missing translation for 'onlineSavingsMsg'
Learn More
Suberoyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11440428

Suberoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11440428
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11440428

Brand: Thermo Scientific Alfa Aesar B24580.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10027-07-3
Molecular Formula C8H12Cl2O2
Molecular Weight (g/mol) 211.082
MDL Number MFCD00000764
InChI Key PUIBKAHUQOOLSW-UHFFFAOYSA-N
Synonym suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichlorideShow MoreShow Less
PubChem CID 534653
IUPAC Name octanedioyl dichloride
SMILES C(CCCC(=O)Cl)CCC(=O)Cl

Specifications

Density 1.172
Boiling Point 162°C to 163°C (15 mmHg)
Flash Point >110°C (230°F)
Linear Formula ClOC(CH2)6COCl
Refractive Index 1.468
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 211.09
Percent Purity 97%
Chemical Name or Material Suberoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251322 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB24580.06,B24580.14
Lot Number10215987 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberB24580,B24580.06,B24580.14
Lot NumberJ7515A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24580.06,B24580.14
Lot Number10215987 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24580.06,B24580.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Suberoyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.