missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Subframe, Adjustable In Heightup To 600 mm, For Un30Pa
Brand: Memmert™ UN30paB29745
This item is not returnable.
View return policy
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30 Series Model | |
Subframe | |
22 in. (Size 110 to 160), 26 in. (Size 30 to 75) | |
560 mm (Size 110 to 160), 660 mm (Size 30 to 75) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Subframe, Adjustable In Heightup To 600 mm, For Un30Pa >
Spot an opportunity for improvement?Share a Content Correction