missing translation for 'onlineSavingsMsg'
Learn More

Melford Succinic Acid

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford S42000-1000.0

Product Code. 30018814

  • £282.00 / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: Butanedioic Acid
  • CAS Number: 110-15-6
  • Molecular Formula: C4H6O4
  • Molecular Weight: 118.1
Specifications

Specifications

Succinic Acid
110-15-6
C4H6O4
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Melford Succinic Acid > 1 kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.