Learn More
Succinic Acid 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 185.0°C to 190.0°C |
Boiling Point | 235.0°C |
Assay Percent Range | 99% |
Linear Formula | HO2CCH2CH2CO2H |
Solubility Information | Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride |
Formula Weight | 118.09 |
Percent Purity | 99% |
Quantity | 250 g |
Flash Point | 206°C |
Packaging | Plastic bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466234 | Certificate of Analysis | 25/09/2024 | 158740100,158740010,158740025,158742500,158745000 |
A0461211 | Certificate of Analysis | 20/03/2024 | 158740100,158740010,158740025,158742500,158745000 |
A0450263 | Certificate of Analysis | 10/02/2023 | 158740100,158740010,158740025,158742500,158745000 |
A0394478 | Certificate of Analysis | 15/12/2022 | 158740100,158740010,158740025,158742500,158745000 |
A0397958 | Certificate of Analysis | 14/12/2022 | 158740100,158740010,158740025,158742500,158745000 |
Safety and Handling

- Succinic acid
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
19Minutes
0Seconds
25New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.