missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, 99%

Product Code. 10573131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10573131

Brand: Thermo Scientific Acros 158740025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 185.0°C to 190.0°C
Boiling Point 235.0°C
Assay Percent Range 99%
Linear Formula HO2CCH2CH2CO2H
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride
Formula Weight 118.09
Percent Purity 99%
Quantity 2.5 kg
Flash Point 206°C
Packaging Plastic bottle
Chemical Name or Material Succinic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.