missing translation for 'onlineSavingsMsg'
Learn More
Succinic acid, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11317588

Succinic acid, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11317588
500 g
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11317588

Brand: Thermo Scientific Alfa Aesar 033272.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry; as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Beilstein 1754069
Merck Index 14,8869
Solubility Information Soluble in water (increasing with temperature); Soluble in ethanol,ethyl ether,acetone and methanol; Insoluble in toluene,benzene,carbon disulfide,carbon tetrachloride and petroleum ether.
Formula Weight 118.09
Percent Purity ≤99.0%
Grade ACS
Density 1.56 g/mL
Quantity 500 g
Flash Point 206°C (402°F)
Physical Form Crystalline
Chemical Name or Material Succinic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU09K003 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number033272.36,033272.A3
Lot NumberW01J035 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog Number033272.36,033272.A3
Lot NumberX31I015 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog Number033272.36,033272.A3
Lot NumberQ09F031 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number033272.36,033272.A3
Lot NumberS22I016 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog Number033272.36,033272.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinic acid, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.