missing translation for 'onlineSavingsMsg'
Learn More
Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals Product Code.: 10215970

Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals

Product Code. 10215970
100 g
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10215970

Brand: Thermo Scientific Acros 158751000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 120.0°C
Color Brown to Yellow
pH 7 to 9 (0.4M soln.)
Loss on Drying 37.0 % to 41.0 % (90°C)
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Quantity 100 g
Beilstein 02, 606
Merck Index 15, 8808
Solubility Information Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol
Formula Weight 270.13
Percent Purity 0.97
Insoluble Matter 0.005% max.
Physical Form Liquid
Chemical Name or Material Succinic acid, disodium salt hexahydrate, 0.97
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468237 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158751000,158750010,158755000
Lot NumberA0442218 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number158751000,158750010,158755000
Lot NumberA0464547 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number158750010,158751000,158755000
Lot NumberA0456667 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number158750010,158751000,158755000
Lot NumberA0452527 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number158750010,158751000,158755000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.