missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, disodium salt hexahydrate, 97%, Thermo Scientific Chemicals

£17.69 - £84.42

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10215970
View Documents
Thermo Scientific Acros
158751000
100 g
£17.69
100g
Please to purchase this item. Need a web account? Register with us today!
10796581
View Documents
Thermo Scientific Acros
158755000
500 g
£51.96
500g
Please to purchase this item. Need a web account? Register with us today!
10708932
View Documents Promotion Details
Thermo Scientific Acros
158750010
1 kg
£84.42
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6106-21-4
270.142
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
CHEBI:63686
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
C4H16Na2O10
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
3083938
disodium;butanedioate;hexahydrate

Specifications

6106-21-4
Brown to Yellow
37.0 % to 41.0 % (90°C)
0.97
C4H16Na2O10
15, 8808
Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
270.142
CHEBI:63686
0.97
Liquid
120.0°C
7 to 9 (0.4M soln.)
Authentic
Glass bottle
02, 606
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium;butanedioate;hexahydrate
3083938
270.13
0.005% max.
Succinic acid, disodium salt hexahydrate, 0.97
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only