missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Acid (White Crystals), Fisher BioReagents
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Description
Succinic Acid is used in the preparation of cell culture and tissue culture media.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 2.7 |
CAS | 110-15-6 |
Melting Point | 185°C |
Boiling Point | 235°C |
Color | White |
Assay Percent Range | ≥99.5 % |
Molecular Formula | C4H6O4 |
Linear Formula | HOOCCH2CH2COOH |
MDL Number | MFCD00002789 |
Merck Index | 15, 8999 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'