missing translation for 'onlineSavingsMsg'
Learn More

Succinic anhydride, 99%

Product Code. 11443960
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
500g
This item is not returnable. View return policy

Product Code. 11443960

Brand: Thermo Scientific Alfa Aesar A12245.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-30-5 | C4H4O3 | 100.07 g/mol

Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

Solubility
Soluble in chloroform.Soluble in chloroform, carbon tetrachloride and alcohol. Slightly soluble in ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00005525
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
PubChem CID 7922
ChEBI CHEBI:36595
IUPAC Name oxolane-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Melting Point 118°C to 121°C
Boiling Point 261°C
Assay Percent Range 0.99
Beilstein 108441
Merck Index 148870
Solubility Information Soluble in chloroform;Soluble in chloroform,carbon tetrachloride and alcohol; Slightly soluble in ether; Insoluble in water.
Formula Weight 100.07
Percent Purity 99%
Density 1.572
Quantity 500 g
Flash Point 157°C (314°F)
Sensitivity Moisture sensitive
Chemical Name or Material Succinic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Succinic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Succinic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.