missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Anhydride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 116.0°C to 120.0°C |
Boiling Point | 261.0°C |
Quantity | 5 kg |
Packaging | Plastic drum |
Assay Percent Range | 99% |
Viscosity | 3 mPa.s (125°C) |
Beilstein | 17,407 |
Fieser | 04,468 |
Merck Index | 15,9 |
Solubility Information | Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C) |
Show More |
Safety and Handling
Product Identifier
- Succinic anhydride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'