Learn More
Succinic Anhydride, 99%, Thermo Scientific Chemicals
Succinic anhydride, 99%, C4H4O3, CAS Number-108-30-5, 2,5-diketotetrahydrofuran, 2,5-furandione, dihydro, butanedioic anhydride, dihydro-2,5-furandione, dihydrofuran-2,5-dione, rikacid sa, succinic acid anhydride, succinic anhydride, succinyl anhydride, succinyl oxide, 1kg, 17,407, 261.0 deg.C
£23.30 - £1265.00
Chemical Identifiers
CAS | 108-30-5 |
---|---|
Molecular Formula | C4H4O3 |
Molecular Weight (g/mol) | 100.07 |
MDL Number | MFCD00005525 |
InChI Key | RINCXYDBBGOEEQ-UHFFFAOYSA-N |
Synonym | succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa |
PubChem CID | 7922 |
ChEBI | CHEBI:36595 |
IUPAC Name | oxolane-2,5-dione |
SMILES | O=C1CCC(=O)O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10744353
|
Thermo Scientific Acros
158760050 |
5 g | Glass Bottle |
£23.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10639803
|
Thermo Scientific Acros
158760500 |
50 g | Plastic bottle |
£32.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10786874
|
Thermo Scientific Acros
158762500 |
250 g | Plastic bottle |
£101.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10417000
|
Thermo Scientific Acros
158760010 |
1 kg | Plastic bottle |
£291.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906171
|
Thermo Scientific Acros
158760051 |
5 kg | Plastic drum |
£1265.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-30-5 | |
100.07 | |
RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
7922 | |
oxolane-2,5-dione |
C4H4O3 | |
MFCD00005525 | |
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
CHEBI:36595 | |
O=C1CCC(=O)O1 |
Specifications
116.0°C to 120.0°C | |
261.0°C | |
C4H4O3 | |
3 mPa.s (125°C) | |
04,468 | |
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
oxolane-2,5-dione | |
7922 | |
100.07 | |
Succinic anhydride |
108-30-5 | |
99% | |
MFCD00005525 | |
17,407 | |
15,9 | |
Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C) | |
O=C1CCC(=O)O1 | |
100.07 | |
CHEBI:36595 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-570-
- Succinic anhydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only