Learn More
Succinimide, 98%, Thermo Scientific Chemicals
£22.10 - £430.14
Chemical Identifiers
CAS | 123-56-8 |
---|---|
Molecular Formula | C4H5NO2 |
Molecular Weight (g/mol) | 99.08 |
MDL Number | MFCD00005495 |
InChI Key | KZNICNPSHKQLFF-UHFFFAOYSA-N |
Synonym | succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione |
PubChem CID | 11439 |
ChEBI | CHEBI:9307 |
IUPAC Name | pyrrolidine-2,5-dione |
SMILES | C1CC(=O)NC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10757582
|
Thermo Scientific Acros
348650050 |
5 g | Glass bottle |
£22.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337371
|
Thermo Scientific Acros
348651000 |
100 g | Glass bottle |
£28.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10738162
|
Thermo Scientific Acros
348655000 |
500 g | Glass bottle |
£58.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13488518
|
Thermo Scientific Acros
348650051 |
5 kg | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-56-8 | |
99.08 | |
KZNICNPSHKQLFF-UHFFFAOYSA-N | |
11439 | |
pyrrolidine-2,5-dione |
C4H5NO2 | |
MFCD00005495 | |
succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
CHEBI:9307 | |
C1CC(=O)NC1=O |
Specifications
123-56-8 | |
Brown to White | |
201°C | |
98% | |
MFCD00005495 | |
15,9001 | |
Solubility in water: 240g/L (20°C). Other solubilities: 1g/24 mL alcohol - 1g/5 mL alcohol (60°C), insoluble in ether and chloroform | |
C1CC(=O)NC1=O | |
99.08 | |
CHEBI:9307 | |
98% | |
Succinimide, 98% |
121.0°C to 126.0°C | |
285.0°C to 290.0°C | |
Authentic | |
C4H5NO2 | |
21,369 | |
succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione | |
KZNICNPSHKQLFF-UHFFFAOYSA-N | |
pyrrolidine-2,5-dione | |
11439 | |
99.08 | |
Powder or Flakes |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-635-6
missing translation for 'rtecsNumber' : WN2200000
missing translation for 'tsca' : TSCA
- Succinimide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only